SP Bel-Art Frigimat® Junior Dry Ice Maker, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F38886-0000
47750-570EA 388.24 USD
47750-570 47750-572 35751-178
SP Bel-Art Frigimat® Junior Dry Ice Maker, Bel-Art Products, a part of SP
Dry Ice Makers
Dry ice flakes are useful for packing around irregularly shaped items and are easily produced as needed with the Frigimat® Junior Dry Ice Maker.
  • Makes 0.77 kg (1.7 lb.) of flakes per bag
  • Useful for packing around irregularly shaped items
  • Produce dry ice easily on demand
  • Shipping weight: 0.54 kg (1.2 lbs)
A brass nozzle screws onto any liquid CO2 cylinder equipped with a dip tube
Makes 0.77kg (1.7 lb) of flakes per bag. The unit cone measures 13 cmL (5") x 5.1 cm diameter (2") at end. The bag is 15W x 30 cmL (6 x 12").
When the cylinder valve is opened about ¾ turn, liquid CO2 expands into the bag to create dry ice snow. The valve is then closed and the bag is ready to be removed and emptied.