VWR® Humidity/Temperature Pen with Memory
Supplier: VWR International
35519-049 10754-542 10754-728
35519-049EA 131.51 USD
35519-049 10754-542 10754-728
VWR® Humidity/Temperature Pen with Memory
Hygrometers Digital Thermo-Hygrometers
Pocket pen is ideal for measuring humidity and temperature quickly and easily.
  • Fast-response, solid-state sensors
  • Traceable®
  • Displays °C and °F
Pen continuously and simultaneously displays relative humidity and temperature
Handy pocket clip makes the pocket pen easy to carry. High-impact ABS plastic case is chemical-resistant.
Meets federal requirements for monitoring temperature and humidity in labs (CLIA specification 493.1252). A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Features dual recall memories that display highest and lowest humidity and temperature readings attained during a designated time period. Pen has fast-response, solid-state sensors that respond in less than 30 seconds. Pen then updates display every second. Temperature is offered in °C or °F. The 1.9 cm (3/4") high LCD display can be easily read at a glance.
Delivery information: Supplied with a lithium battery (CR2032, 3 V) for continuous, always-on monitoring for half a year.