Anti-CD325 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 8C11]

Supplier: Biolegend
350809 350810
350809-BLEA 158.8 USD
350809-BL 350810-BL
Anti-CD325 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 8C11]
Antibodies
Alexa Fluor® 488 anti-human CD325 (N-Cadherin) [8C11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD325
Clonality: Monoclonal
Clone: 8C11
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR