Anti-Ki-67 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: Ki-67]

Supplier: Biolegend
350507 350508 350532
350507-BLEA 153.1 USD
350507-BL 350508-BL 10758-982
Anti-Ki-67 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: Ki-67]
Antibodies
Alexa Fluor® 488 anti-human Ki-67 [Ki-67]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Bovine; Apps: ICFC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by our Ki-67 staining protocol below. For flow cytometric staining using the μg size, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. For flow cytometric staining using test sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. For immunofluorescence microscopy, a concentration range of 1-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ki-67
Clonality: Monoclonal
Clone: Ki-67
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR