Anti-CD294 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BM16]

Supplier: Biolegend
350107 350108
350107-BLEA 105.8 USD
350107-BL 350108-BL
Anti-CD294 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BM16]
Antibodies
FITC anti-human CD294 (CRTH2) [BM16]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD294
Clonality: Monoclonal
Clone: BM16
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR