Barbituric acid ≥98%, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine

Management of Change (MOC) category = N

B654-05
35001-242EA 180.11 USD
35001-242
Barbituric acid ≥98%, J.T.Baker®
Barbituric acid
Formula: C₄H₄N₂O₃
MW: 128.09 g/mol
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 250…252 °C
Density: 0.95 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006666
CAS Number: 67-52-7
EINECS: 200-658-0
Merck Index: 13,00966

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Loss on Drying at 105°C ≤2 %
Solubility (4% in H₂O) Passes Test
Assay (C₄H₄N₂O₃) (by acid-base titration) ≥98 %
Appearance Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR