Anti-CD243 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UIC2]

Supplier: Biolegend
348607 348608
348607-BLEA 167.8 USD
348607-BL 348608-BL
Anti-CD243 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UIC2]
Antibodies
APC anti-human CD243 (MDR-1) [UIC2]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD243
Clonality: Monoclonal
Clone: UIC2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR