Anti-CD317 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: RS38E]

Supplier: Biolegend
348405 348406
348405-BLEA 175.1 USD
348405-BL 348406-BL
Anti-CD317 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: RS38E]
Antibodies
PE anti-human CD317 (BST2, Tetherin) [RS38E]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Pigtail, Rhesus Macaque; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD317
Clonality: Monoclonal
Clone: RS38E
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Pigtail;Rhesus Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR