TouchNTuff® 92-600 Disposable Nitrile Gloves with Enhanced Chemical Splash Protection
Supplier: Ansell Healthcare
585836 585834 585835 585837 92600060 92600110
34800-504CS 441.93 USD
34800-504 34800-500 34800-502 34800-506 77587-114 77587-116
TouchNTuff® 92-600 Disposable Nitrile Gloves with Enhanced Chemical Splash Protection
Gloves
An unparalleled combination of chemical splash protection, comfort, and durability.
  • TNT™ Technology for ultimate protection and advanced chemical resistance
  • Extra-soft, smooth feel, boosting comfort for extended wear
  • Proprietary nitrile resists rips and tears to provide outstanding durability
TouchNTuff® 92-600 gloves are the world's leading disposable gloves for chemical splash protection. Made with TNT™ chemical splash resistance technology, these gloves effectively safeguard hands from exposure to harmful chemicals, with a super-soft nitrile consturction that provides a comfortable feel during extended wear. Proprietary formulation is extra-strong to prevent tearing during heavy-duty work. The silicone-free design prevents contaminant transfer that causes flaws in paints, coatings, and finishes.
Running on 59% renewable energy recycling min. 14% of water to reduce freshwater usage. Certified by Intertek for diverting >99% of waste from landfill. With accredited environmental management system. Plastic-free, recyclable packaging includes min. 70% recycled material.
Certifications: Category III. EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016. FDA21 CFR 177-2600. US Food Contact Approved. ISO 18889:2019, ISO 9001, UKCA.
Caution: Do not use against heat or cold. Do not use for mechanical nor extended chemical protection.