VWR® Specific Gravity Hydrometer Set
Supplier: VWR International
34630-000
34630-000EA 687.93 USD
34630-000
VWR® Specific Gravity Hydrometer Set
Hydrometers
This Hydrometer Set consists of eight DURAC® Specific Gravity/Relative Density (g/cmᶟ) Plain Form Hydrometers for measuring specific gravity of heavy and light liquids in various ranges from 0.700 to 2.000
  • Hydrometers are 300 mm (12”) long and made with lead free ballast and glass
  • Temperature standardization: 60 °F (15 °C)
  • Thermometer is 300 mm (12”) long and made of lead-free glass; contains standard mercury-free blue organic liquid fill against a lead-free white back glass
  • All units are individually serialized
  • Hand inspected by our U.S. quality team
Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
Set also includes one general-purpose liquid-in-glass thermometer to reference liquid temperature for accurate hydrometer readings.
Packaging: Packaged in a deluxe storage case with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Caution: Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.