VWR® Specific Gravity/Relative Density Plain Form Hydrometers, Traceable to NIST
Supplier: VWR International
34615-006 34620-006 34620-100 34620-176 34620-180 34622-001 34622-103 34622-158 34622-205 34622-250 34623-150 34625-600 34629-601 34610-002 34620-156 34620-158 34620-162 34620-164 34620-166 34620-168 34620-170 34620-172 34620-174 34620-178 34623-106 34623-208 34623-252 34623-300 34625-633 34625-666 34629-011 61801-4711
34615-006EA 53.95 USD
34615-006 34620-006 34620-100 34620-176 34620-180 34622-001 34622-103 34622-158 34622-205 34622-250 34623-150 34625-600 34629-601 34610-002 34620-156 34620-158 34620-162 34620-164 34620-166 34620-168 34620-170 34620-172 34620-174 34620-178 34623-106 34623-208 34623-252 34623-300 34625-633 34625-666 34629-011 30617-754
VWR® Specific Gravity/Relative Density Plain Form Hydrometers, Traceable to NIST
Hydrometers
These plain form hydrometers are available in a variety of models measuring specific gravity/relative density (g/cm³) of liquids lighter and heavier than water in various ranges from 0.690 to 3.000.
  • Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60 °F (15 °C)
Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
Certifications: Hand inspected by our U.S. quality team, individually serialized.
Packaging: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.