Anti-CD271 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: ME20.4]

Supplier: Biolegend
345109 345110
345109-BLEA 179 USD
345109-BL 345110-BL
Anti-CD271 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: ME20.4]
Antibodies
PE/Cy7 anti-human CD271 (NGFR) [ME20.4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD271
Clonality: Monoclonal
Clone: ME20.4
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR