Anti-BTLA Mouse Monoclonal Antibody [clone: MIH26]

Supplier: Biolegend
344502
344502-BLEA 329.9 USD
344502-BL
Anti-BTLA Mouse Monoclonal Antibody [clone: MIH26]
Antibodies
Purified anti-human CD272 (BTLA) [MIH26]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: BTLA
Clonality: Monoclonal
Clone: MIH26
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR