Orion™ ISE Calibration Standards, Thermo Scientific
Supplier: Thermo Fisher Scientific
841109 930707 040908 940916 040906 040907 951006 951207 943506 922006 941706 941708 941707 923206 940906 940907 945306 948206 920706 920707 951007 921906 941107 841108 941105 948207 950206 950207 977007 942906 650700 F40904 F40905
34104-914EA 129.05 USD
34104-914 34117-330 34182-623 34182-645 34182-667 34182-689 34183-080 34183-091 34183-138 34183-182 34184-119 34184-120 34184-141 34184-301 34184-356 34184-389 34184-469 34184-538 34184-607 34184-618 34184-673 34184-753 34184-833 34184-877 34184-888 34184-902 34184-050 34184-083 34184-174 34184-200 14223-216 76070-176 76070-178
Orion™ ISE Calibration Standards, Thermo Scientific
Standards Environmental Analysis Standards
Take reproducible ISE measurements with Orion™ ISE Calibration Standards
  • Orion calibration standards are NIST traceable.
  • Orion calibration standards are manufactured under ISO 9001 quality standards.
Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.
Use the Orion ISE calibration standards with our extensive line of ion selective electrodes (ISEs) for ammonia, ammonium, bromide, cadmium, calcium, carbon dioxide, chloride, chlorine, copper, cyanide, fluoride, iodide, lead, nitrate, potassium, silver, sodium and sulfide analysis. Whether you are in the lab or in the field, you can be confident that Orion solutions will deliver accurate and repeatable measurements. Orion solutions are produced in controlled batches using ultrapure water and undergo rigorous quality testing throughout the process to prevent contamination. Our strict quality procedures verify correct bottle labeling and volume, so each batch meets specific lot release criteria. Bottles are sealed and torque-tested to ensure bottle caps are secure and solutions remain free of contamination. Rigorous lifetime testing ensures sealed Orion solutions remain within published specifications for their entire shelf life and do not degrade over time.
Ordering information: The Fluoride standard bulk pack includes 4x475mL each of 1ppm fluoride standard premixed with TISAB II and 10ppm fluoride standard premixed with TISAB II.
Packaging: All standards are supplied in 1 pint (475 mL) bottles, unless otherwise noted