Buffer Assortment Pack, Hydrion Buffer Chemvelopes, Micro Essential Lab
Supplier: MICRO ESSENTIAL LABORATORY I
280-2-11
34176-085EA 62.83 USD
34176-085
Buffer Assortment Pack, Hydrion Buffer Chemvelopes, Micro Essential Lab
Buffers pH Calibration Buffers
Buffer assortment pack includes buffers with pH ranges from 2 to 11. Chemvelopes are made of a triple protective foil consisting of aluminum, polyethylene and paper which keeps the Hydrion Buffers dry and free-flowing.
Hydrion buffer chemvelopes produce 500 ml of pH buffer standard solution when dissolved in distilled water.
All buffers are also available separately.