Certified pH Buffer Set
Supplier: MICRO ESSENTIAL LABORATORY I
CC-471V
34175-485EA
42.17
USD
34175-485
Certified pH Buffer Set
Buffers
pH Calibration Buffers
Set includes calibrated and pre-weighed amounts of buffer powders at three pH values (pH 4.0, 7.0, and 10.0) for testing or calibrating your pH meter.
Powders come in capsules and each is sufficient for 100 ml of buffer solution.
Delivery information: Includes 10 capsules for each pH value and a vial of buffer preservative.