Wright's stain, powder, HARLECO® stain for hematology, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

376-29
34172-114EA 3699 USD
34172-114 34172-076 15204-110
Wright's stain, powder, HARLECO® stain for hematology, Sigma-Aldrich®
Wright's stain

For differential blood staining.


  • For In-Vitro Diagnostic Use.
  • For Professional Use.


Certified by the Biological Stain Commission (BSC).

Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
EINECS: 273-541-5
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR