Wright's stain pack para Hema-Tek® Stainer, HARLECO®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

65043-93
34172-112EA 199 USD
34172-112
Wright's stain pack para Hema-Tek® Stainer, HARLECO®, Sigma-Aldrich®
Wright's stain

HARLECO® Designed for use with the Ames Hema-Tek® Stainer


  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use

Flash Pt: 15.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR