Safranine O, HARLECO®, certified by the Biological Stain Commission, Sigma-Aldrich®

Supplier: MilliporeSigma

350-12
34172-064EA 193 USD
34172-064 11214-844
Safranine O, HARLECO®, certified by the Biological Stain Commission, Sigma-Aldrich®
Safranine O

Used in various staining protocols such as Gram staining. Can be used as a cytological counterstain.

Formula: C₂₀H₁₉ClN₄
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR