Leishman's eosin-methylene blue, powder, HARLECO® stain for hematology, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Eosin and methylene blue Leishman, Eosin-Methylene blue dye Leishman, Leishman's stain, Leishman stain, Leishman's stain (eosin methylene blue compound)

252-12
34172-032EA 331 USD
34172-032
Leishman's eosin-methylene blue, powder, HARLECO® stain for hematology, Sigma-Aldrich®
Leishman's Eosin-Methylene blue

Used for blood, malarial parasites and bone marrow staining


Certified by the Biological Stain Commission (BSC)

MDL Number: MFCD00131498
CAS Number: 12627-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR