5-Sulfosalicylic acid solution 3%, HARLECO® for microbiology, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-Hydroxy-5-sulphobenzoic acid, 2-Hydroxy-5-sulfobenzoic acid

1506A-75
34172-026EA 99 USD
34172-026
5-Sulfosalicylic acid solution 3%, HARLECO® for microbiology, Sigma-Aldrich®
5-Sulfosalicylic acid solutions

Used for semi-quantitative visual determination of albumin in urine by SSA precipitation method

Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00007508
CAS Number: 5965-83-3

Order Now

Specification Test Results

N/A N/A

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR