Alcohol 90%, HARLECO® for histology, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

65348-85
34172-022CS 365 USD
34172-022
Alcohol 90%, HARLECO® for histology, Sigma-Aldrich®
Alcohol

Part of the QSM Histology stain line. Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols.

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Expiration date 18 months from mfg date
Color (APHA) 10 APHA max
Form Clear liquid
Identity (IR-spectrum) Conforms to standard
Specific Gravity at 25/25°C 0.801-0.817


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR