Alcohol 100%, HARLECO® for histology, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

65347-85
34172-020CS 300 USD
34172-020
Alcohol 100%, HARLECO® for histology, Sigma-Aldrich®
Alcohol

Part of the QSM Histology stain line. Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols.

Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Expiration date 18 months from mfg date
Assay (Ethanol) 89.0-92.0%
Assay (Isopropyl alcohol) 4.0-6.0%
Assay (Methanol) 3.5-5.5%
Color (APHA) 10APHA max
Form Clear liquid
Infrared Spectrum Conforms to standard
Residue after evaporation 0.001% max
Water (H2O) 0.075% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR