Orion™ Sure-Flow® Reference Electrode, Ag/AgCl Single Junction Half-Cell, Thermo Scientific
Supplier: Thermo Fisher Scientific
900100
34107-923EA 321.39 USD
34107-923
Orion™ Sure-Flow® Reference Electrode, Ag/AgCl Single Junction Half-Cell, Thermo Scientific
Electrodes pH Electrodes
This reference half-cell electrode helps to precisely measure with ion-selective electrodes. Ideal for ISE determinations and routine pH measurements with Orion™ 910100 electrodes.
  • Ag/AgCl reference
  • Sure-Flow® sleeve junction
Reference electrode is recommended for all pH and ion-selective measurements.
For these samples, the double junction reference electrode, Orion 900200, with an appropriate intermediate filling solution, is recommended.
When filled with Orion™ filling solution 900001 (34111-830), this electrode matches the potential characteristics of a conventional KCl saturated calomel electrode.