VWR® pH Electrode Arm
Supplier: VWR International
34105-859 470204-504
34105-859EA 286.86 USD
34105-859 470204-504
VWR® pH Electrode Arm
Electrode Holders
Electrode arm moves in almost any direction, holding up to three electrodes safely and securely in selected positions.
  • Ideal for multiple pH readings
  • Accepts all standard electrodes
  • Pivots 360°, Extends 53 cm (207/8")
The arm articulates at the three points to maintain electrodes in vertical position.
Designed to accept any standard glass, reference, combination, or other electrode — even a temperature compensation probe. Weighted base and spring counter-balance permit smooth, fluid movement with superior stability.