Corning® High Volume Stirrer, Corning
Supplier: Corning
6795-611
34103-633CS 1382.67 USD
34103-633
Corning® High Volume Stirrer, Corning
Stirrers Magnetic Stirrers
Ideal for stirring large volumes of liquid
Designed for 19L (5 gal.) vessels, but can stir up to 38L (10 gal.). Compact design conserves bench space. Stirrer uses a magnet made of rare earth metals for exceptional coupling strength that helps center the stir bar quickly inside the vessel. Heavy-duty motor and rugged ABS plastic top make unit long-lasting. Top size is 279x279 mm (11x11").
Ordering information: Fluoropolymer resin-coated stir bar available separately.