VORTEX 3 Orbital Shakers, IKA® Works
Supplier: IKA Works
0003340001
33993-874EA 1005.32 USD
33993-874
VORTEX 3 Orbital Shakers, IKA® Works
Shakers and Mixers Orbital Shakers
Orbital shakers are suitable for touch-operated or short-time continuous operation
Shaker attachments and inserts securely click onto units in any position and allow various applications to be performed. The one-hand attachment is 88mm (37/16") in diameter for larger vessels. The universal attachment allows use of various inserts for tubes, flasks, and plates.
Self-ventilating motor is encased in a sturdy zinc die-cast housing. Compact shakers feature wide, infinitely adjustable speed ranges. Special silicone base feet with ultra-high vibration damping ensure unit stability at the highest speeds.
Ordering information: Supplied with one standard attachment for test tubes and small vessels.