Barnstead/Thermolyne Explosion-Proof Hot Plate, Safe-T HP6, Thermo Scientific
Supplier: Thermo Fisher Scientific
HP11515B
33922-655EA 2023.5 USD
33922-655
Barnstead/Thermolyne Explosion-Proof Hot Plate, Safe-T HP6, Thermo Scientific
Heaters Hot Plates
Designed for Class I, Group D hazardous areas
Explosion-proof. “Limit” thermostat cuts off unit when temperature reaches 243°C (469.4°F). Sealed aluminum housing contains and protects controls. Cast aluminum top plate ensures maximum heat transfer. Corrosion-resistant steel case is easily cleaned and maintained. Integral thermostatic control maintains top plate temperature ±5.5°C (±9.9°F). Accommodates up to 11.3kg (25lb.) loads.
Ordering information: Integral mounting lugs, connection for 1.27cm (1/2") conduit and 45.7cm (18") lead wire included for safe installation. Conduit installation required. For optional dial thermometer, see 33922-677.