Barnstead/Thermolyne Electric Hot Plate, Type 2300, Thermo Scientific
Supplier: Thermo Fisher Scientific
HP2305BQ
33922-305EA 401.11 USD
33922-305
Barnstead/Thermolyne Electric Hot Plate, Type 2300, Thermo Scientific
Heaters Hot Plates
For laboratories where an inexpensive heat source is needed and bench space is scarce.
Ruggedly built to withstand continuous usage. Cast aluminum heating surface reaches a top temperature of 371°C (700°F) in less than 71/2 minutes. The nickel-chromium heating element is embedded in special heat-conducting refractory material for maximum protection. Stepless, thermostatic control allows selection of temperature from approximately 38° to 371°C (100° to 700°F). Perforated stainless steel case.
Ordering information: Complete with 1.5m (5') three-wire cord and plug.