PowrTwin Power Controller, Glas-Col®
Supplier: Glas-Col
104APL1202
33797-440EA 678.82 USD
33797-440
PowrTwin Power Controller, Glas-Col®
Controllers Temperature Controllers
For precise manual control of heating tapes, cords, and mantles
Supplied with lighted on/off switch, three-wire load receptacle, and three-wire line cord with molded plug.
Solid state, proportional voltage controller is capable of controlling two separate circuits independently. Output range adjustable from 3–100% of rated voltage. Can be used on bench top or can be mounted on frame using clamp provided on back panel.