Holder for Rugged Rotator, 75 to 200 mm Test Tubes, Glas-Col
Supplier: Glas-Col
099ARD50
33797-310EA 786.81 USD
33797-310
Holder for Rugged Rotator, 75 to 200 mm Test Tubes, Glas-Col
Support Stands
Double duty holder for bottles and flasks.
  • Load balanced by easy adjustment
  • Retaining plate lined to protect glassware
Used with the Rugged Rotator and holds a complete rack of stoppered test tubes for rotating end over end. Will hold rack 12×5".
Ordering information: Holder only.
Delivery information: 7 to 10 days after order.