Cole-Parmer® HM-200 Series Multiposition Extraction Heaters, Antylia Scientific
Supplier: Argos Technologies
03076-03 03076-24 03076-27 03076-06 03076-33 03076-09 03076-30 03076-00
33749-200EA 2046.28 USD
33749-200 33749-299 33749-324 33749-233 33749-380 33749-266 33749-357 33749-175
Cole-Parmer® HM-200 Series Multiposition Extraction Heaters, Antylia Scientific
Heaters Heating Mantles
Allows liquids with different boiling points to be heated next to each other.
  • Unique vented airflow ensures the case remains 'cool-to-the-touch'
  • Heat solutions simultaneously in each position
  • Robust construction permits continuous operation of the mantles as required by routine extraction tests
Regulating the heat on these multiposition extraction mantles is simple using the built-in solid-state electronic controls. The units accept round-bottom flasks ranging from 100 to 1000 ml. Select an extraction heater depending on the size of your flask.
Extraction heaters feature an independent heater per mantle, allowing liquids with different boiling points to be heated alongside each other. Each heater has its own control dial and on/off indicator light, and can reach up to a maximum element temperature of 450 °C (842 °F).
The control panel is safely separated from the heating elements by a well-ventilated air space. A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater. Powder-coated aluminum surfaces and polypropylene top cover ensure good chemical resistance and easy cleanup.
Certifications: CE marked.
Delivery information: Supplied with two 28" long rods, one 55" long rod, support rod clamps, and 6' (1.8 m) power cord with US plug.