Cole-Parmer® CN-200 Series Analog Heating Controllers, Antylia Scientific
Supplier: Antyila Scientific
36223-02 36223-03 36223-05
33743-384EA 389.81 USD
33743-384 33743-386 76045-894
Cole-Parmer® CN-200 Series Analog Heating Controllers, Antylia Scientific
Controllers Temperature Controllers
Cost-effective control of resistive heating loads.
  • Provide manual control of uncontrolled heating mantles, tapes, and cords
  • Compact—use on benchtop or mount to a frame
Use these external one-way controllers to manually regulate the power input to laboratory heating equipment. Each heating controller features two indicators: 'Power On' white neon light and 'Mantle/Bunsen Heater On' amber neon light. As you turn the control dial clockwise to increase power, the amber indicator light begins to pulsate. Once maximum power is reached, the light will glow continually.
Heating controllers come with a short mains lead with an IEC socket for connecting to the resistive load. Easily place on benchtop or mount on a 0.5" (12.5 mm) diameter rod using the included support rod clamp.
The heating controllers deliver different levels of power according to your requirements and are suitable for EM series Electromantles, the Electric Bunsen and Heating Cords and Tapes.
Delivery information: Supplied with support clamp at rear for standard 12.5 mm dia rod (not included) and power cord.