Lab Clamp, Glas-Col®, LLC
Supplier: Glas-Col
099DA283040
33730-280EA 123.23 USD
33730-280
Lab Clamp, Glas-Col®, LLC
Clamps General Purpose Clamps
Right angle clamp.
  • Sturdy construction
  • Easily attached
  • Easy adjustment
Our right angle clamp is ideal for light duty use on 3/4" rods.
Delivery information: 7-10 days after order.