Shaker with 180° Shaking Motion, Glas-Col

Supplier: Glas-Col
099AWS18012
33725-050EA 3023.54 USD
33725-050
Shaker with 180° Shaking Motion, Glas-Col
Shakers and Mixers Rotating/Rocking Shakers
Simulates hand shaking by rotating through 180° arc.

  • Shakes two flasks simutaneously
  • Bench top mounting
  • Use with screw cap or side arm flasks

For use with small to medium peptide synthesis flasks and culture flasks (up to 2-¹/₄" diameter, 10" long). This shaker offers 180 degree agitation shaking for solid phase peptide synthesis and for hard to dissolve solids such as polymers.

Ordering information: Shaker base with 2 large finger clamps.

Delivery information: 7 to 10 days after order.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR