Shaker with 180° Shaking Motion, 240 V

Supplier: Glas-Col
099AWS18024
33725-048EA 3145.53 USD
33725-048
Shaker with 180° Shaking Motion, 240 V
Shakers and Mixers Rotating/Rocking Shakers
Simulates hand shaking by rotating through 180° arc.

  • 240 V model
  • Shakes two flasks simutaneously
  • Bench top mounting
  • Use with screw cap or side arm flasks

For use with small to medium peptide synthesis flasks and culture flasks (up to 2-¹/₄" diameter, 10" long). This shaker offers 180 degree agitation shaking for solid phase peptide synthesis and for hard to dissolve solids such as polymers.

Ordering information: Shaker base with 2 large finger clamps.

Delivery information: 7 to 10 days after order.

Caution: 240 V model. Corresponding model with voltage suitable for use in US (33725-050) is also available, please see separate product page.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR