Tissue Culture Disk Head for Mini Rotator, Glas-Col
Supplier: Glas-Col
099ATC132
33725-046EA 561.51 USD
33725-046
Tissue Culture Disk Head for Mini Rotator, Glas-Col
Shakers and Mixers Rotating/Rocking Shakers
Aluminum disk with 30 position clips for 8 mm to 14 mm tubes.
  • Designed for quick change
  • Large and small micro centrifuge and micro test tube applications
Interchangeable disk for use with the mini rotator which quickly changes to increase your capabilities.
Ordering information: Tissue culture disk head with spring clamps.