Safety ECO Funnels
Supplier: CP Lab Safety
EF-8-JUSTRITE-B2-SYS EF-8-JUSTRITE-B5-SYS EF-8-JUSTRITE-B2-SYS-HPLC EF-8-FS70 EF-8-FS70-HPLC EF-4-38-SYS EF-4-53B-SYS EF-4-38 EF-8-83B EF-4717-1C EF-3004C EF-3004C-HPLC EF-4-53B EF-30020 EF-4716-1P EF-3008 EF-8-JUSTRITE-B EF-4-38-65-SYS EF-4-38-82C-SYS EF-4-38-85C-SYS EF-4-38-85-SYS EF-4-38GL2-SYS EF-4-38GLSC-SYS EF-4-38GL-SYS EF-4-38-SYS-HPLC EF-4-38Y EF-4-38YGL2-SYS EF-4-FS70C-SYS EF-4-FS70C-SYSB EF-4-FS70-SYS EF-4-FS70-SYSB EF-4-FS70Y EF-4-JUSTRITE-B2-SYS EF-4-JUSTRITE-B5CR-SYS EF-4-JUSTRITE-B5C-SYS EF-4-JUSTRITE-B5R-SYS EF-4-JUSTRITE-B5-SYS EF-8-FS70G-SYS EF-8-FS70G-SYSB EF-8-FS70G-SYS-HPLC EF-8-FS70-SYS EF-8-FS70-SYSB EF-8-FS70Y EF-8-FS70Y-HPLC EF-8-FS70Y-SYS EF-8-FS70Y-SYSB EF-8-FS70Y-SYS-HPLC EF2-8-FS70CG-SYS EF2-8-FS70C-SYS EF2-8-FS70CY-SYS EF2-8-FS70G EF2-8-FS70G-SYS EF2-8-FS70-SYS EF2-8-FS70Y EF2-8-FS70Y-SYS EF2-8-SW EF2-8-SWB EF-4-38YGLSC-SYS EF-4-38YGL-SYS EF-4-38Y-HPLC EF-4-38Y-SYS EF-4-38Y-SYS-HPLC EF-4-63B EF-4-63B-SYS EF-4-63BY EF-4-63BY-SYS EF-4-63C-SYS EF2-8-83BB-SYS EF2-8-83BG EF2-8-83BG-SYS EF2-8-83B-SYS EF2-8-83BY EF2-8-83BY-SYS EF2-8-FS70 EF2-8-FS70B EF2-8-FS70B-SYS EF2-8-FS70CB-SYS EF-3009Y EF-3009Y-HPLC EF-3009Y-SYS EF-4-30020 EF-4-30020C-SYS EF-4-30020-SYS EF-4-3009 EF-4-3009C-SYS EF-4-3009CY-SYS EF-8-83C5-SYS-TELLI EF-8-83CB-SYS EF-8-83CG-SYS EF-8-83C-SYS EF-8-83CY-SYS EF-8-FS70B EF-8-FS70B-HPLC EF-8-FS70B-SYS EF-8-FS70B-SYSB EF-8-FS70B-SYS-HPLC EF-30020B EF-30020B-SYS EF-30020CB-SYS EF-30020CG-SYS EF-30020C-SYS EF-30020CY-SYS EF-30020G EF-30020G-SYS EF-30020-SYS EF-30020Y EF-3004CY-SYS EF-3004GLB-SYS EF-3004GLG-SYS EF-3004GLSC-SYS EF-3004GLSCY-SYS EF-3004GL-SYS-TELLI EF-3004GLY-SYS EF-3008B EF-3008B-SYS EF-3009B-HPLC EF-3009B-SYS EF-3009CB-SYS EF-3009CG-SYS EF-3009C-SYS EF-3009CY-SYS EF-3009G EF-3009G-HPLC EF-3009G-SYS EF-3009-SYS EF-4-JUSTRITE-BY EF-4-JUSTRITE-BY2-SYS EF-4-JUSTRITE-BY5C-SYS EF-4-JUSTRITE-BY5-SYS EF-4-SF-029 EF-4-SW EF-4-SWC-SYS EF-4-SWCY-SYS EF-4-SW-SYS EF-4-SWY EF2-8-83B EF2-8-83BB EF-4717-1CG EF-4717-1CG-SYS EF-4717-1C-SYS EF-4717-1CY EF-4717-1CY-SYS EF-4-83B EF-4-83B-SYS EF-4-83BY EF-4-83C-SYS EF-4-FS70 EF-8-JUSTRITE-B2B-SYS EF-8-JUSTRITE-B2B-SYS-HPLC EF-8-JUSTRITE-B2G-SYS EF-8-JUSTRITE-B2G-SYS-HPLC EF-8-JUSTRITE-B2Y-SYS EF-8-JUSTRITE-B2Y-SYS-HPLC EF-8-JUSTRITE-B5BC-SYS EF-8-JUSTRITE-B5C-SYS EF-8-JUSTRITE-B5GC-SYS EF-8-JUSTRITE-BB EF-8-SWCG-SYS EF-8-SWC-SYS EF-8-SWCY-SYS EF-8-SWG EF-8-SWG-SYS EF-8-SW-SYS EF-8-SWY EF-8-SWY-SYS EF-4-3009-SYS EF-4-3009Y EF-4-3009Y-SYS EF-4-38-006N EF-4-38-008N EF-4-38-400 EF-4-38-4004-SYS EF-4-38-400-SYS EF-4-38-62C-SYS EF-4-38-65C-SYS EF-4-63CY-SYS EF-4716-1PB EF-4716-1PB-SYS EF-4716-1PG EF-4716-1PG-SYS EF-4716-1P-SYS EF-4716-1PY EF-4716-1PY-SYS EF-4717-1CB EF-4717-1CB-SYS EF-8-JUSTRITE-BB5-SYS EF-8-JUSTRITE-BG EF-8-JUSTRITE-BG5-SYS EF-8-JUSTRITE-BY EF-8-JUSTRITE-BY5C-SYS EF-8-JUSTRITE-BY5-SYS EF-8-SW EF-8-SWB EF-8-SWB-SYS EF-8-SWCB-SYS EF2-8-SWB-SYS EF2-8-SWCG-SYS EF2-8-SWC-SYS EF2-8-SWCY-SYS EF2-8-SWG EF2-8-SWG-SYS EF2-8-SW-SYS EF2-8-SWY EF2-8-SWY-SYS EF-30020Y-SYS EF-3004CB EF-3004CB-HPLC EF-3004CB-SYS EF-3004CG EF-3004CG-HPLC EF-3004CG-SYS EF-3004C-SYS-TELLI EF-3004CY EF-3004CY-HPLC EF-3008CB-SYS EF-3008CG-SYS EF-3008C-SYS EF-3008CY-SYS EF-3008G EF-3008G-SYS EF-3008-SYS EF-3008Y EF-3008Y-SYS EF-3009B EF-4-SWY-SYS EF-8-83B5-SYS EF-8-83BB EF-8-83BB-SYS EF-8-83BG EF-8-83BG-SYS EF-8-83B-SYS EF-8-83BY EF-8-83BY-SYS EF-8-83C5-SYS EF-8-FS70CB-SYS EF-8-FS70CB-SYSB EF-8-FS70CG-SYS EF-8-FS70CG-SYSB EF-8-FS70C-SYS EF-8-FS70C-SYSB EF-8-FS70CY-SYS EF-8-FS70CY-SYSB EF-8-FS70G EF-8-FS70G-HPLC
33500-974EA 1020.31 USD
33500-974 33500-980 33500-988 33501-836 33501-838 33501-840 33501-844 47735-342 47735-346 47735-370 30246-504 30246-506 47735-344 47735-362 47735-366 47735-378 33500-976 77676-894 77676-896 77676-898 77676-900 77676-902 77676-904 77676-906 77676-908 77676-910 77676-912 77676-974 77676-976 77676-978 77676-980 77676-982 77676-984 77676-986 77676-988 77676-990 77676-992 77677-074 77677-076 77677-078 77677-080 77677-082 77677-084 77677-086 77677-088 77677-090 77677-092 77676-714 77676-716 77676-718 77676-720 77676-722 77676-724 77676-726 77676-728 77676-730 77676-732 77676-914 77676-916 77676-918 77676-920 77676-922 77676-924 77676-926 77676-928 77676-930 77676-932 77676-694 77676-696 77676-698 77676-700 77676-702 77676-704 77676-706 77676-708 77676-710 77676-712 77676-854 77676-856 77676-858 77676-862 77676-864 77676-866 77676-868 77676-870 77676-872 77677-034 77677-036 77677-038 77677-040 77677-042 77677-044 77677-046 77677-048 77677-050 77677-052 77676-754 77676-756 77676-758 77676-760 77676-762 77676-764 77676-766 77676-768 77676-770 77676-772 77676-794 77676-796 77676-798 77676-800 77676-802 77676-806 77676-808 77676-810 77676-812 77676-834 77676-836 77676-838 77676-840 77676-842 77676-844 77676-846 77676-848 77676-850 77676-852 77676-994 77676-996 77676-998 77677-000 77677-002 77677-004 77677-006 77677-008 77677-010 77677-012 77676-690 77676-692 77676-954 77676-956 77676-958 77676-960 77676-962 77676-964 77676-966 77676-968 77676-970 77676-972 77677-094 77677-096 77677-098 77677-100 77677-102 77677-104 77677-106 77677-108 77677-110 77677-112 77677-134 77677-136 77677-138 77677-140 77677-142 77677-144 77677-146 77677-148 77676-874 77676-876 77676-878 77676-880 77676-882 77676-884 77676-886 77676-888 77676-890 77676-892 77676-934 77676-936 77676-938 77676-940 77676-942 77676-944 77676-946 77676-948 77676-950 77676-952 77677-114 77677-116 77677-118 77677-120 77677-122 77677-124 77677-126 77677-128 77677-130 77677-132 77676-734 77676-738 77676-740 77676-742 77676-744 77676-746 77676-748 77676-750 77676-752 77676-774 77676-776 77676-778 77676-780 77676-782 77676-784 77676-786 77676-788 77676-790 77676-792 77676-814 77676-816 77676-818 77676-820 77676-822 77676-824 77676-826 77676-828 77676-830 77676-832 77677-014 77677-016 77677-018 77677-020 77677-022 77677-024 77677-026 77677-028 77677-030 77677-032 77677-054 77677-056 77677-058 77677-060 77677-062 77677-064 77677-066 77677-068 77677-070 77677-072
Safety ECO Funnels
Funnels Safety Funnels
ECO Funnel® is the ultimate lab safety product for collection of hazardous liquid waste. When securely attached onto an open waste container, it prevents 99.9% of toxic fumes from being released into lab. ECO Funnel is designed to prevent fires caused by ignited vapors as well as spills of hazardous waste from open containers, which helps laboratories stay compliant with local health and safety codes.
  • Latching lid with gasket to keep fumes contained
  • Removable large particle filter catches stir bars or other items
  • Detachable tube extends into waste container to minimize chemical evaporation
The larger 8" version of ECO Funnel includes an additional safety feature to prevent over-filling; a secondary air-lock tube creates a small air gap in your waste container, which causes fluid to back up temporarily into the funnel when the container is full. After the screw cap adapter is loosened, the remaining fluid will safely flow into the container and not spill.
Ordering information: ECO Funnels can be ordered individually or as a complete system. Systems include an ECO Funnel and corresponding waste container. Some systems also include a secondary container option for maximum safety and compliance.