Adapter, Rotary Evaporator Bump Trap, R200 and R205, Chemglass
Supplier: Chemglass
CG-1317-B-10
33501-708EA 398.83 USD
33501-708
Adapter, Rotary Evaporator Bump Trap, R200 and R205, Chemglass
Adapters Adapters, Joints with Tube Connectors
Vapor tube/bump trap adapter is designed to be used with the Buchi® R-200/205 rotary evaporators.
  • This adapter combines a vapor tube adapter and a bump trap
  • Tapered sidewalls permit continuous washing with condensed solvent
  • While preventing vapor cooling points along the path of the condenser
  • The upper stem has two large holes flush with the lower end
  • Allows solvent vapor to flow freely
  • This portion of the trap is easy to clean
Adapters have a 50 mm outer Ø top flange with a tapered interior to fit the Büchi® sealing ring. Lower portion of the adapter has the listed standard taper inner joint.
Note: Items will replace the Buchi® part numbers as listed. Chemglass items are to be used with the Keck clip, not the Buchi Combi-clip.
Delivery information: Each adapter is supplied complete with the corresponding size keck clip.