EasySat™ Bucketless Floor Mops, Contec®
Supplier: Contec
BLMOP BLVKMOP BLVKMOPIR TBLVKMOP TBLVKMOPIR BLMOPQC
33501-534EA 493.11 USD
33501-534 33501-536 33501-538 33501-540 33501-542 76239-666 76239-980 76377-432
EasySat™ Bucketless Floor Mops, Contec®
Mops
Designed specifically for cleanroom use, this mop system completely eliminates the need for bucket or wringer.
  • Hydrophilic polyester/clean ester, laser cut sealed edge foam material
  • Translucent tank allows user to see solution
  • Fully autoclavable for critical areas
  • Easy to use
  • Compatible with a wide range of solvents
System includes a 1.5 l (52 oz.) translucent solution tank that can clean up to 5,000 square feet. Tank weighs only 3.2 kg (7 lbs.) when full. A push button flow control releases cleaning solution on demand. The angled, 55.9 cm (22") mop heads clean hard-to-reach areas and can be attached and removed within seconds.
System and mop heads are fully autoclavable up to 121 °C (250 °F) for 30 minutes. Mop heads come in regular and textured styles and are also available gamma irradiated.
It features lasercut sealed-edge VertiKlean® mop head material, which is a lightweight polyester fabric laminated to clean ester foam for excellent abrasion resistance.
Ordering information: Mop system and mop heads must be purchased separately.