Support Stands, Stainless Steel, Chemglass
Supplier: Chemglass
CG-1539-S-100
33501-428EA 1651.81 USD
33501-428
Support Stands, Stainless Steel, Chemglass
Supports Flask Supports
Floor style stainless steel support stands.
  • Made to accept 12 and 22 liter CG-1539 half jacketed flasks
  • Top of stand has a 39" reinforced riser to support overhead stirrers, clamps and so on
Stand is 67" overall height with a depth of 22" ring has a diameter of 81/2 " I.D. and is 28" above floor level.