VWR® Traceable® Key Chain Micro IR Thermometer
Supplier: VWR International
33501-416
33501-416EA 90.85 USD
33501-416
VWR® Traceable® Key Chain Micro IR Thermometer
Thermometers Digital Thermometers
Pocket-size, economical, infrared thermometer captures temperature readings of any surface — liquid, solid, and semi-solid — in less than a second.
  • Traceable®
  • Easy and non-invasive operation
  • Reads °C or °F
  • Pocket-sized
Operation is easy and non-invasive. Simply turn on the thermometer, point it at the sample, and take the reading. The no-touch temperature measurements are ideal for applications such as food preparation, life science, field use, cleanroom, and fast inspections. Features include an easy-to-read 1.3 cm (1/2") display, soft-touch keys, clock, key chain, low battery alert, and battery with 40 hours of continuous life. Thermometer reads in °C or °F.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.