Casework, Laminate, Base Cabinet Trim Parts, Removable Access Panels
Supplier: Case Systems
R2200300136 R2200300148 R2200360136 R2200360148
33501-180EA 229.38 USD
33501-180 33501-182 33501-184 33501-186
Casework, Laminate, Base Cabinet Trim Parts, Removable Access Panels
Case Systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
  • Plastic laminate provides superior resistance to chemicals
  • Scratch and abrasion resistant
The key component of Case Systems casework is the plastic laminate. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring while eliminating sharp edges and corners.
The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards. Construction features an industrial-grade, environmentally responsible WOODSTALK® engineered board core from Dow BioProducts, using formaldehyde-free resin to bind wheat-straw fiber, and heavy-duty institutional grade hardware. The result — cabinets that are as attractive as they are durable.
Case cabinetry gives you the adaptability necessary in today’s laboratories. The unique hardware system provides rigid support for vertical integration without limiting adjustability. Vertical supports are slotted, allowing infinite height adjustment of overhead cabinets, electrical raceways, services, and reagent shelves. Color: Beige.
Ordering information: Case Systems laminate laboratory furniture has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor Furniture Specialist.