Polypropylene Bucket System, Contec®
Supplier: Contec
2720
33501-018EA 444.65 USD
33501-018
Polypropylene Bucket System, Contec®
Buckets
Fully autoclavable polypropylene rolling cart with two 6.5 gallon (25 l) buckets and wringer.
  • Durable plastic bucket system is lightweight and easy to maneuver
  • System includes two different color buckets for easy solution identification
  • Fully autoclavable system is ideal for sterile environment use
Durable, yet lightweight construction makes the 2720 bucket system easy to maneuver around cleanrooms. Red and blue buckets allow for differentiation between clean and waste solutions. Ideal for use with Contec's Edgeless® Mop.