Bottle, Narrow Mouth, with Screw Cap, CP Lab Safety
Supplier: CP Lab Safety
EC-2004
33500-972EA 3 USD
33500-972
Bottle, Narrow Mouth, with Screw Cap, CP Lab Safety
Bottles Plastic Bottles
4 Liter narrow-mouth LDPE bottle with 38 mm polypropylene screw cap.
  • Unlined closure
  • 38-430 neck finish
  • Natural colour