KoolTemp® GTS 6-Panel EPS Insulated Container, Cold Chain Technologies
Supplier: Cold Chain Technologies
131-2-UK
33500-330CS 550.6 USD
33500-330
KoolTemp® GTS 6-Panel EPS Insulated Container, Cold Chain Technologies
Mailers and Shippers
Six-panel sheet insulated containers provide space-saving, durable, and efficient thermal protection for virtually any temperature-sensitive application.
  • Designed for transporting and shipping temperature-sensitive articles
  • Step-by-step assembly instructions included
  • Kit does not include refrigerants
Container kit includes a polyethylene liner, which provides additional leakage protection for any payload. Containers meet or exceed standards of all major shipping carriers.