Thermocouple Adaptor, PTFE, Chemglass
Supplier: Chemglass
CG-1048-A-01 CG-1048-A-02 CG-1048-A-03 CG-1048-A-04 CG-1048-A-05 CG-1048-A-06 CG-1048-A-07
33500-232EA 80.37 USD
33500-232 33500-234 33500-236 33500-238 33500-240 33500-242 33500-244
Thermocouple Adaptor, PTFE, Chemglass
Adapters Adapters, Joints with Tube Connectors
PTFE adapter for use with either 1/8” or 1/4” thermocouples.
  • Design permits use with CG-145 Keck clips
  • Outer Viton® o-ring
  • Forms an airtight seal
Adapter has an outer Viton® o-ring to form a grease-free, airtight seal between the adapter and the listed size of outer joint. A compression nut works in conjunction with the upper Viton® to provide a airtight seal between the adapter and thermocouple.