Anti-CD30 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BY88]

Supplier: Biolegend
333904
333904-BLEA 231.9 USD
333904-BL
Anti-CD30 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BY88]
Antibodies
FITC anti-human CD30 [BY88]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: BY88
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR