CleanTex™ Foam Covered Cotton Swab, with Wood Shaft, Advantus
Supplier: Advantus
CT700A
33344-046PK 291.25 USD
33344-046
CleanTex™ Foam Covered Cotton Swab, with Wood Shaft, Advantus
Swabs Swabs for Cleaning
A general-purpose swab for cleaning surfaces and hard-to-reach areas.
  • Polyurethane highly absorbent foam head surrounding a cotton bud core, hermetically sealed around the bud
  • Open cell head possesses excellent capacity and cushioning, while internal cotton bud provides cushioned support and absorbency
  • Soft and non abrasive, silicone-free
  • Handles lubricants with no degeneration
  • Tip thickness: 10.1 mm (0.39")
The long hardwood handle allows the head to access recessed areas. Use to clean with solvents such as IPA. Apply lubricants and other liquids and remove excess materials.