Uvex Futura™ Goggles, Honeywell Safety
Supplier: Honeywell Safety Products
S345C
33002-179CS 905.62 USD
33002-179
Uvex Futura™ Goggles, Honeywell Safety
Goggles
Wrap-around, sports-style goggles provide excellent panoramic vision. They feature a frosted brow guard to reduce glare and indirect venting to increase airflow.
  • Flexible PVC body, textured neoprene headband
  • Impact-resistant polycarbonate lens protects against dust, mist, and splashing
  • Clear lenses are coated with Uvex­treme® lens coating for anti-fog, anti-scratch, anti-static, and anti-UV protection
  • Fit over most prescription eyeglasses
The goggle body is made of flexible PVC material for increased comfort. Headband is made of textured neoprene, appropriate for chemical applications. Goggles fit over most prescription eyeglasses. Lenses can be easily replaced.
Certifications: Meet ANSI Z87+ and CSA Z94.3.